Learn More
Emma Safety Footwear Venus S3 Safety Shoes
Product Code. 15998891
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15998891 43 1 pair
15918891 35 1 pair
15928891 36 1 pair
15938891 37 1 pair
15948891 38 1 pair
15958891 39 1 pair
15968891 40 1 pair
15978891 41 1 pair
15988891 42 1 pair
15908901 44 1 pair
15918901 45 1 pair
15928901 46 1 pair
15938901 47 1 pair
15948901 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15998891 Supplier Emma Safety Footwear Supplier No. MM71454643
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The Venus is a low profile, easy on / easy off shoe. The smooth, easy to clean contours offer additional hygiene advantages over and above lace up shoes. It's perfect for a 'quick change' from office to factory floor or walking between one facility and another.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Easy Twist
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Elastic
Color Black
Design Venus S3
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 43
Toe Material Steel