missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VEPH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110567
This item is not returnable.
View return policy
Description
Immunogen sequence: KSLAFHTMLT KGVGSDDGED ENRGDIPASI SLSEIDPLGQ GNDKLPFKTD TERSQLGESS VSYPNIIHID SENLSETVKE NSQEETPETT ASPIEYQDKL.
Ventricular zone-expressed PH domain-containing protein homolog 1 (VEPH1), also known as KIAA1692 or Protein melted, is a 833 amino acid protein belonging to the MELT/VEPH family. VEPH1 is a peripheral membrane protein localized to the cytoplasmic side. VEPH1 has one PH domain which is required for the membrane targeting of the protein. The human gene encoding VEPH1 is 76% homologous to the Drosophila VEPH1 gene which encodes a protein that acts as a regulator of the insulin signaling pathway. It is thought that this role of VEPH1 is conserved across species. VEPH1 exists as three isoforms produced by alternative splicing.
Specifications
| VEPH1 | |
| Polyclonal | |
| Unconjugated | |
| VEPH1 | |
| 2810471M23Rik; KIAA1692; MELT; Protein melted; protein melted homolog; RGD1308217; ventricular zone expressed PH domain containing 1; ventricular zone expressed PH domain homolog 1; ventricular zone expressed PH domain-containing 1; ventricular zone-expressed PH domain-containing protein 1; Ventricular zone-expressed PH domain-containing protein homolog 1; VEPH; VEPH1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 72789, 79674 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.68 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A1A535, Q14D04 | |
| VEPH1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 448-547 of human VEPH1 (NP_078897.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction