Learn More
Usbeck Laborgeraete Verbandschere, Edelstahl 18/8, L=130mm,
Brand:  Usbeck Laborgeraete 4240
Verbandschere, Edelstahl 18/8, L=130mm,spitz-stumpf
Product Code. 10669902
  • ONLINE EXCLUSIVE PRICE
    € 7.00
    € 6.12 / Each
    Save € 0.88
    12% Off
Only null left
Estimated Shipment: 09-01-2026