missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verenigde Kunststof Bedrijven™ HDPE, Alveolit Sealring, Screw Closure DIN 51 (Red)
Click to view available options
Color:
Red
Description:
Screwcap DIN 51
Description
- HDPE screw cap
- leak-proof closure
- Applicable on plastic packages
Specifications
Product Type | Screw Closure |
Description | Screwcap DIN 51 |
Material | HDPE, Alveolit |
Color | Red |
For Use With (Equipment) | Bottle and Jerrican |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'