Learn More
3M™ Versaflo™ V–500E Air Regulator
Product Code. 11387270
Change view
Click to view available options
Power Intensity:
3.5 - 8 bar Working Pressure
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Power Intensity unitSize
11387270 3.5 - 8 bar Working Pressure Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11387270 Supplier 3M™ Supplier No. 7100005806
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
For Use With (Application) Regulator ensures an individually-adjustable, consistently controlled airflow
Power Intensity 3.5 - 8 bar Working Pressure
Product Type V–500E Air Regulator
Quantity 1 Ea.