missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H7N1 Antibody (InA334), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-50816DL594
This item is not returnable.
View return policy
Description
Influenza A H7N1 Monoclonal antibody specifically detects Influenza A H7N1 in Virus samples. It is validated for ELISA, ELISA, Direct ELISA
Specifications
| Influenza A H7N1 | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/Netherlands/219/03 (strain H7N7). | |
| 0.25 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| ELISA, Sandwich ELISA, Direct ELISA | |
| InA334 | |
| ELISA, Sandwich ELISA, Direct ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction