missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Versican Recombinant Rabbit Monoclonal Antibody (24GB2380)

Rabbit Recombinant Monoclonal Antibody
€ 198.00 - € 564.00
Specifications
| Antigen | Versican |
|---|---|
| Clone | 24GB2380 |
| Concentration | 0.4 mg/mL |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275759
![]() |
Invitrogen™
MA557840 |
100 μL |
€ 564.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274786
|
Invitrogen™
MA557841 |
20 μL |
€ 198.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Versican Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Versican is a member of the family of large aggregating proteoglycans (also including aggrecan, brevican, and neurocan). The protein shows wide tissue distribution which includes fibrous, articular, and elastic cartilages, as well as nervous, epidermal, arterial, and loose connective tissues. Versican V1 has been shown to enhance cell proliferation, and also induce cell transformation and protect cells from apoptosis. This was demonstrated through its ability to downregulate the expression of proapoptotic Bad. Versican's existence as a extracellular matrix protein in human airways has also lead to the discovery of an inflammatory role in asthmatics.Specifications
| Versican | |
| 0.4 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P13611, Q62059, Q9ERB4 | |
| 114122, 13003, 1462 | |
| A synthesized peptide derived from human Versican (aa 3200-3280). | |
| Antibody |
| 24GB2380 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| VCAN | |
| 5430420N07Rik; 9430051N09; Chondroitin sulfate proteoglycan 2; chondroitin sulfate proteoglycan 2 (versican); chondroitin sulfate proteoglycan core protein 2; Cspg2; DPEAAE; ERVR; GHAP; glial hyaluronate-binding protein; hdf; heart defect; large fibroblast proteoglycan; NG2; PGM; PG-M; PG-M core protein; PG-M(V0); PG-M(V1); V1 Neo; VCAN; versican; versican core protein; Versican core protein precursor (Large fibroblast proteoglycan) (Chondroitin sulfate proteoglycan core protein 2) (PG-M); versican proteoglycan; Versican V0; WGN; WGN 1; WGN1 | |
| VCAN | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title