Learn More
Saint Gobain Life Sciences Versilic™ Silicone Caps
Use these stoppers for biology, microbiology and hematology, as well as food science, technical, medical and hospital apparatus. Versilic™ Silicone Caps are particularly suited for all types of tissue culture work.
Brand: Saint Gobain Life Sciences 263107
Additional Details : Weight : 0.80000kg
Description
- Biologically neutral and non-toxic silicone caps for closing tubes and flasks
- Odorless, tasteless, non-adherent and resistant to aging
- Nontoxic, biologically neutral
- FDA/BGA approved
- For test tubes
- Easy to install
Specifications
Silicone Stopper | |
107 mm | |
Silicone | |
Autoclavable |
94 mm | |
500 Pk. | |
-70 to +250°C | |
FDA Compliant Food Contact, BFR, BGA |