missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Vertigo Check and Go Black PU3 Gloves

Catalog Number 15700908
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Protect your hands when handling sharp objects in a dry or slightly greasy environment. Honeywell Safety Products™ Vertigo Check and Go Black PU3 Gloves are knitted to provide a conforming, snug fit and good dexterity.

  • Knitted glove for snug conformation to the hand.
  • Spectra™ Lycra™ thread construction for comfort, cut resistance, and dirt resistance.
  • Polyurethane-coated palm for abrasion resistance.
  • Good dexterity.
  • Ventilated back for breathability.
  • Sturdy.
  • Not for heat exposure.
  • Large cut resistance value marking (3) on the glove back for easy identification.
TRUSTED_SUSTAINABILITY
Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standard, EN 388-2003 Mechanical risks, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/11/08/0443 EXT 02/10/2015, EC Attestation Number 0075/797/162/11/08/0443 EXT 02/10/2015
Classification Dexterity: 5
Coated Area Palm, Fingers
Description Gauge 13
Coating Material Polyurethane
Color Black
Cut Resistance Rating 3
Material Lycra, Spectra Fiber
Product Type Gloves
Size 11
For Use With (Application) Automotive, Aviation, Building, Construction, Catering, Energy, Electricity, Glass, Logistics, Maintenance, Metal, Paper, Service, Telecom, Transportation and Utility industries.
Puncture Rating 2
Quantity 20/PK
Tear Rating 4
Length (Metric) 23 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'