missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Vertigo Check and Go Black PU3 Gloves

Catalog Number 15750898
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Protect your hands when handling sharp objects in a dry or slightly greasy environment. Honeywell Safety Products™ Vertigo Check and Go Black PU3 Gloves are knitted to provide a conforming, snug fit and good dexterity.

  • Knitted glove for snug conformation to the hand.
  • Spectra™ Lycra™ thread construction for comfort, cut resistance, and dirt resistance.
  • Polyurethane-coated palm for abrasion resistance.
  • Good dexterity.
  • Ventilated back for breathability.
  • Sturdy.
  • Not for heat exposure.
  • Large cut resistance value marking (3) on the glove back for easy identification.
TRUSTED_SUSTAINABILITY
Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-2003 EEC Basic standard, EN 388-2003 Mechanical risks, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/11/08/0443 EXT 02/10/2015, EC Attestation Number 0075/797/162/11/08/0443 EXT 02/10/2015
Classification Dexterity: 5
Coated Area Palm, Fingers
Description Gauge 13
Coating Material Polyurethane
Color Black
Cut Resistance Rating 3
Material Lycra, Spectra Fiber
Product Type Gloves
Size 6
For Use With (Application) Automotive, Aviation, Building, Construction, Catering, Energy, Electricity, Glass, Logistics, Maintenance, Metal, Paper, Service, Telecom, Transportation and Utility industries.
Puncture Rating 2
Quantity 20/PK
Tear Rating 4
Length (Metric) 23 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'