missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human RNase L ELISA Kit (Colorimetric)

Product Code. 30239501 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30239501

Brand: Novus Biologicals™ NBP340377

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Colorimetric ELISA Kit

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.625 - 1000 pg/mL (example only; lot dependent)
Assay Sensitivity 7.3 pg/mL (example only; lot dependent)
Detection Method Colorimetric
Immunoassay Kit Format Pre-coated 96T Strip Plate
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Tissue Homogenates, Other Biological Fluids
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 6041
Gene Symbol RNASEL
Quantity 1 Kit
Research Discipline Chromatin Research, Cytokine Research, DNA replication Transcription Translation and Splicing, Immunology, Innate Immunity, Prostate Cancer, Protein Kinase
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym 2-5A-dependent ribonuclease, 2-5A-dependent RNase, 2'-5'-oligoisoadenylate synthetase-dependent, EC 3.1.26.-, HPC1, interferon-induced 2-5A-dependent RNase, MGC104972, MGC133329, PRCA1, prostate cancer 1, Ribonuclease 4, Ribonuclease L, ribonuclease L (2'-5'-oligoisoadenylate synthetase-dependent), RNase L, RNS4DKFZp781D08126
Target Human
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.