Learn More
EVOS™ Vessel Holders and Stage Plates
EVOS™ vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides.
Merk:  EVOS™ AMEPVH020
Overige gegevens : Gewicht : 0.50000kg
 Bekijk meerdere types van dit product via de
Produktcode. 15441074
  • € 162.00 / 1 stuk
Geschatte verzenddatum
bij leverancier 26-07-2019
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
Product Features:
  • EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment.
  • EVOS stage plates hold vessel holders, thereby expanding the number of vessel holders available for the EVOS systems.
  • With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Spezifikation
Spezifikation
EVOS™ FL Auto Imaging System
Vessel Holder for Automated Stage, one Terasaki plate
1 holder
EVOS vessel holder; Dimensions: 128mm x 86mm; Capacity: 1 Terasaki plate
Videos
Dokumentation