missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VEZF1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15544642
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15544642 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15544642 Supplier Invitrogen™ Supplier No. PA541131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: SCNICGKLLS AAYITSHLKT HGQSQSINCN TCKQGISKTC MSEETSNQKQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

VEZF1 is a possible transcription factor. It specifically binds to the CT/GC-rich region of the interleukin-3 promoter and mediates tax transactivation of IL-3.Transcriptional regulatory proteins containing tandemly repeated zinc finger domains are thought to be involved in both normal and abnormal cellular proliferation and differentiation. ZNF161 is a C2H2-type zinc finger protein. See MIM 603971 for general information on zinc finger proteins.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen VEZF1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene VEZF1
Gene Accession No. Q14119
Gene Alias AI848691; AW046909; DB1; putative transcription factor DB1; Vascular endothelial zinc finger 1; VEZF1; Zinc finger protein 161; ZNF161
Gene Symbols VEZF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human VEZF1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7716
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.