missing translation for 'onlineSavingsMsg'
Learn More
Learn More
vFLIP Antibody (6/14), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-78138CL1
This item is not returnable.
View return policy
Description
vFLIP Monoclonal antibody specifically detects vFLIP in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| vFLIP | |
| Monoclonal | |
| CoraFluor 1 | |
| Rat | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 2024-06-14 00:00:00 | |
| PBS | |
| Full-length recombinant vFLIP expressed in Escherichia coli | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction