missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGCNL1 Antibody (S187-7), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22410IR
This item is not returnable.
View return policy
Description
VGCNL1 Monoclonal specifically detects VGCNL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| VGCNL1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NALCN | |
| Fusion protein amino acids 1659-1738 (cytoplasmic C-terminus) of rat NALCN | |
| 0.1 mL | |
| 259232 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| S187-7 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| bA430M15.1, CanIonFLJ44659, FLJ23913, FLJ44764, four repeat voltage-gated ion channel, MGC74524, sodium leak channel non-selective protein, sodium leak channel, non-selective, VGCNL1, voltage gated channel like 1, Voltage gated channel-like protein 1 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects approx 200kDa. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction