missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ VGF Antibody (07) [Alexa Fluor™ 405], Novus Biologicals™

Product Code. 30064721 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30064721 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30064721 Supplier Novus Biologicals™ Supplier No. NBP326899AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VGF Monoclonal antibody specifically detects VGF in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen VGF
Applications ELISA
Classification Monoclonal
Clone 07
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias neuro-endocrine specific protein VGF, neurosecretory protein VGF, VGF nerve growth factor inducible
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VGF (Uniprot#: O15240; aa 1-615).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Endocrinology, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7425
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.