missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VGF Antibody (08) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26900IR
This item is not returnable.
View return policy
Description
VGF Monoclonal antibody specifically detects VGF in Human samples. It is validated for ELISA
Specifications
| VGF | |
| Monoclonal | |
| DyLight 755 | |
| neuro-endocrine specific protein VGF, neurosecretory protein VGF, VGF nerve growth factor inducible | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VGF (Uniprot#: O15240; aa 1-615). | |
| 0.1 mL | |
| Cell Biology, Endocrinology, Neuroscience, Signal Transduction | |
| 7425 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 08 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction