missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VGF Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76906DL594
This item is not returnable.
View return policy
Description
VGF Polyclonal antibody specifically detects VGF in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| VGF | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| neuro-endocrine specific protein VGF, neurosecretory protein VGF, VGF nerve growth factor inducible | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human VGF. The immunogen is located within the last 50 amino acids of VGF. Amino Acid Squence: ARRAQEEAEAEERRLQE | |
| 0.1 mL | |
| Cell Biology, Endocrinology, Neuroscience, Signal Transduction | |
| 7425 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction