missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VGLL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545925
This item is not returnable.
View return policy
Description
Peptide sequence: HSGCAAPGPA SYRRPPSAAT TCDPVVEEHF RRSLGKNYKE PEPAPNSVSI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 86%.
Transcription cofactor vestigial-like protein 4 (VGLL4) acts as a specific coactivator for the mammalian TEFs.
Specifications
| VGLL4 | |
| Polyclonal | |
| Unconjugated | |
| Vgll4 | |
| BC048841; KIAA0121; Transcription cofactor vestigial-like protein 4; vestigial like 4; vestigial like 4 (Drosophila); vestigial like family member 4; Vestigial-like 4; vestigial-like family member 4; VGL-4; Vgll4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9686 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q14135 | |
| Vgll4 | |
| Synthetic peptide directed towards the middle region of human VGLL4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction