Learn More
VGLUT1 Antibody (CL2754) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-46627AF488
Product Code. 30305610
  • € 737.00 / 0.10mL
Description
VGLUT1 Monoclonal antibody specifically detects VGLUT1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VGLUT1
Monoclonal
Alexa Fluor 488
BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1
This antibody was developed using a recombinant protein derived from Q9P2U7, with the exact immunogen sequence remaining proprietary.
0.1 mL
ABC Transporters, Cellular Signaling, Neuronal Cell Markers, Neuroscience
57030
Store at 4⪚C in the dark.
IgG2b
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL2754
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse, Rat