Learn More
Invitrogen™ VGLUT1 Monoclonal Antibody (N28/9), APC
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545503
Product Code. 17827332
  • € 455.00 / 100µg
Description
1 μg/mL of MA5-45503 was sufficient for detection of VGLut1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 52kDa. No cross-reactivity against VGlut2. This antibody was formerly sold as clone S28-9.
A Na(+)-dependent inorganic phosphate cotransporter; may be involved in neuronal Na+ transport.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VGLUT1
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4
Q3TXX4, Q62634, Q9P2U7
SLC17A7
Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1
Immunohistochemistry, Western Blot, Western Blot
N28/9
APC
SLC17A7
2900052E22Rik; AI851913; BNPI; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; brain-specific Na-dependent inorganic phosphate cotransporter; SLC17A1; SLC17A1/VGLUT1; Slc17a7; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 7; solute carrier family 17 (vesicular glutamate transporter), member 7; solute carrier family 17 member 7; Vesicular glutamate transporter 1; VGlU1; VGLUT 1; VGLUT1
Mouse
Protein G
RUO
116638, 57030, 72961
4°C