Learn More
VGLUT2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29209-1-AP-150UL
 View more versions of this product
Product Code. 18056375
  • € 410.00 / 150µL
Description
The VGLUT2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human VGLUT2. This antibody recognizes human, mouse antigen. The VGLUT2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
VGLUT2
600 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, Immunofluorescence, Immunohistochemistry, ELISA
DNPI, SLC17A6, VGLUT2
150 μL
57084
Store at -20°C. Stable for one year after shipment.