Learn More
Invitrogen™ VGLUT3 Monoclonal Antibody (N34/34), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545514
Description
1 μg/mL of MA5-45514 was sufficient for detection of VGLUT3 in 20 μg of CV-1 fibroblast cells (lysate) transfected with VGLUT3 by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 65kDa. This antibody was formerly sold as clone S34-34.
Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-3 is detectable in a few normal cell type spermatids in testis with active spermatogenesis, placental trophoblast membranes, and neurons in brain. GLUT-3 staining is also detectable in human cancers including those of the ovary, lung, and testis.
Specifications
| VGLUT3 | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4 | |
| Q7TSF2, Q8BFU8, Q8NDX2 | |
| SLC17A8 | |
| Fusion protein amino acids 546-588 (cytoplasmic C-terminus) of rat VGLUT3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| N34/34 | |
| APC | |
| SLC17A8 | |
| BC042593; DFNA25; SLC17A8; solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8; solute carrier family 17 (vesicular glutamate transporter), member 8; solute carrier family 17 member 8; Vesicular glutamate transporter 3; vesicular glutamate transporter-3; VGLUT 3; VGLUT3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 216227, 246213, 266767 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.