Learn More
Tocris Bioscience™ VH 032 amide-PEG6-amine
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
VH 032 amide-PEG6-amine is a functionalized von-Hippel-Lindau protein ligand (VHL) for PROTAC® research and development; incorporates an E3 ligase ligand plus a PEG6 linker with terminal amine ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | (2 S,4 R)-1-(-23-Amino-2-(tert-butyl)-4-oxo-6,9,12,15,18,21-hexaoxa-3-azatricosanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C36H57N5O10S0.2 HCl |
| Purity | 0.95 |