missing translation for 'onlineSavingsMsg'
Learn More

VIAAT/SLC32A1/VGAT Antibody (CL2793) [Janelia Fluor™ 549], Novus Biologicals™

Product Code. 30523076 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30523076 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30523076 Supplier Novus Biologicals Supplier No. NBP343787JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VIAAT/SLC32A1/VGAT Monoclonal antibody specifically detects VIAAT/SLC32A1/VGAT in Human,Mouse,Rat samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen VIAAT/SLC32A1/VGAT
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL2793
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias bA122O1.1, hVIAAT, solute carrier family 32 (GABA vesicular transporter), member 1, Solute carrier family 32 member 1, Vesicular GABA transporter, vesicular inhibitory amino acid transporter, VGATGABA and glycine transporter, VIAAT
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of Q9H598, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 140679
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.