Learn More
VICI Jour Chromatography™ Tubing Clip
Product Code. 12955001
Change view
Click to view available options
:
For holding both 1/6 in. and 1/8 in. polymer tubes in correct placement in beakers, flasks, bottles and other labe equipment up to 4mm wall thickness with stainless steel spring
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12955001 For holding both 1/6 in. and 1/8 in. polymer tubes in correct placement in beakers, flasks, bottles and other labe equipment up to 4mm wall thickness with stainless steel spring
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12955001 Supplier VICI Jour Chromatography™ Supplier No. JR9001
Only null left
Specifications
For Use With (Application) holding both 1/6 in. and 1/8 in. polymer tubes in correct placement in beakers, flasks, bottles and other labe equipment up to 4mm wall thickness with stainless steel spring
Product Type Tubing Clip