Learn More
Kern Sohn Video Microscope OIV 255
Description
- The Kern OIV 255 is a video microscope which has been constructed to optimise digital stereo microscopy. Our well-conceived, comprehensive solution with axial optical unit enables immediate, simple display of your samples on the screen
- The LED incident illumination unit (ring) included as standard guarantees the very best illumination of your sample
- Combined with the large working surface, recording objects on the screen is ideally suited for monitoring, analysis and documentation in industrial environments
- The excellent optical unit enables continuous sharp image tracking across the entire zoom range from 0.7x-5x
- The powerful 2.0 megapixel camera of the microscope without eyepieces offers, thanks to the HDMI output, smooth live monitoring of your samples from the HD monitor. In addition, the software which is easy to use, the USB stick as well as the USB mouse which are integral components of the delivery, mean you can process and store your results digitally
- The OIV 255 guarantees software-controlled taking of images and videos with additional, extensive measuring functions
- A protective dust cover, as well as multi-lingual user instructions are included in the scope of the delivery
Specifications
Specifications
| Type | Video Microscope |
| Connector Types | HDMI (60 FPS) |
| Description | Image and video recording and measurements software function |
| Display Type | LED |
| Field of View | 29.8 to 4.2 mm |
| Magnification Power | 0.7X to 5X |
| Resolution | 2 MP |
| Sensor Types | CMOS 1/2 in. |
| Voltage | 230 V |