Learn More
Saline-Sodium Citrate (SSC), 20X Solution (Molecular Biology), Fisher BioReagents
Click to view available options
Quantity:
1L
4L
20L
Unit Size:
1 L
20 L
4 L
Description
Saline–Sodium Citrate (SSC) is used as a buffer in Southern transfer protocols.
Specifications
Specifications
| Chemical Name or Material | Saline-Sodium Citrate |
| Concentration | 2.9 to 3.1 M |
| pH | 7 |
| Physical Form | Liquid |
| Quantity | 1L |
| DNase | DNase free |
| Grade | Molecular Biology |