Learn More
VITLAB™ Polypropylene Test Tube Racks
Specifications
Specifications
| Color | White |
| Holds | Tubes (18 mm) |
| Material | Polypropylene |
| Positions | 55 |
| Stackable | Yes |
| Quantity | 5 Pack |
| Temperature (Metric) Range | -20°C to +90°C |
| Diameter (Metric) Opening | 18 mm |
| Product Type | Test Tube Rack |
| Array | 5 x 11 |