Learn More
S Murray™ Oval Wash Bottles
Product Code. 12881434
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
12881434 250 mL Pack of 5
12891434 500 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12881434 Supplier S Murray™ Supplier No. L780/01
Only null left
Add to basket
Add to basket
Use these wash bottles for a variety of applications. S Murray™ Oval Wash Bottles require only slight pressure to dispense liquid. For general purpose use.
  • Sold in packs of 5
  • Narrow screw neck
Specifications
Capacity (Metric) 250 mL
For Use With (Application) General purpose
Material MDPE
No. per Pack 5 Pack
Width (Metric) 65mm
Closure Color Natural
Closure Included Yes
Shape Oval
Height (Metric) 160 mm
Product Type Wash Bottle