Learn More
Trajan Scientific and Medical™ PerkinElmer Split / Splitless Tapered FocusLiner
Product Code. 12934031
Click to view available options
Quantity:
1 Ea.
25/Pk.
Unit Size:
Each
Pack of 25
Product Code. 12934031
Brand: Trajan Scientific and Medical™ 09209501
Only null left
Add to basket
Add to basket
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with PerkinElmer instruments
Specifications
Quantity 1 Ea.
Length (Metric) 92 mm
Product Type GC Inlet Liner
Diameter (Metric) Inner 4 mm
Diameter (Metric) Outer 6.2 mm