Learn More
Anisomycin, 97%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J62964.MF
| Quantity | 50mg |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- A protein synthesis inhibitor
- Causes apoptosis of PC12 Cells
Specifications
| 22862-76-6 | |
| 50mg | |
| MFCD00077650 | |
| 20705 | |
| (2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)-3-pyrrolidinyl acetate; Flagecidin | |
| YKJYKKNCCRKFSL-BFHYXJOUSA-N | |
| (2S,3R,4R)-4-hydroxy-2-[(4-methoxyphenyl)methyl]pyrrolidin-3-yl acetate | |
| 253602 | |
| 265.31 | |
| Anisomycin |
| White | |
| C14H19NO4 | |
| UN3462 | |
| 14,670 | |
| Soluble in water at 2mg/ml,in DMSO at 20mg/ml,and in methanol at 20mg/ml | |
| COC1=CC=C(C[C@@H]2NC[C@@H](O)[C@@H]2OC(C)=O)C=C1 | |
| 265.31 | |
| CHEBI:338412 | |
| 97% |