Learn More
Retsch™ Test Sieves with 25 mm height
Shop All Retsch Products
Click to view available options
Pore Size:
63 μm
1 mm
100 μm
1.25 mm
125 μm
150 μm
1.70 mm
2.00 mm
250 μm
8.00 mm
800 μm
16.00 mm
Unit Size:
Each
Description
- High quality joining technology guarantees permanently taut wire mesh
- One-piece cast sieve frame and seamless mesh joint prevent cross contamination
- Stainless steel sieve frame with high form stability
- High degree of corrosion resistance and easy cleaning thanks to high-alloy stainless steel
- Excellent product quality due to extensive optical inspection
- Maximum stability and optimum sealing when used in sieve stacks thanks to the o-ring which is placed in the recess designed for this purpose
- Clear and precise labeling of the sieves with full traceability based on individualized laser engraving
Specifications
Specifications
| Pore Size | 150 μm |
| Type | Test Sieve |
| Certifications/Compliance | DIN ISO 3310-1 |
| Material | Stainless Steel |
| Includes | Calibration Certificate |
| Height (Metric) | 25 mm |
| Diameter (Metric) Outer | 200 mm |