Learn More
Bel-Art™ Stainless-Steel Spoons
Product Code. 15560122 Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
0.5 mL, 2 mL
1.6 mL, 3 mL
2 mL, 8 mL
Length (Metric):
15 cm
21 cm
30 cm
Unit Size:
Pack of 2
Product Code. 15560122
Brand: Bel-Art™ H367290021
Only null left
Add to basket
Add to basket
Double-ended spoons offer two bowl sizes for convenient sample handling
  • Polished 304 stainless-steel construction
  • Solid, rounded shaft for easy handling
Specifications
Autoclavable Autoclavable
Capacity (Metric) 1.6 mL, 3 mL
Color Silver
Disposable Reusable
Product Type Spoon
Length (English) 8.26 in.
Includes Double end with 1.6mL small spoon at one end and 3mL large spoon at other
Length (English) Handle 5.8 in.
Length (Metric) Handle 14.73 cm
Material Stainless Steel
Length (Metric) 21 cm