Learn More
Axygen™ FDSS™ Robotic Tips
Product Code. 15581383 Shop All Axygen Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Hamamatsu FDSS 7000 System
Unit Size:
Pack of 19200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15581383 Hamamatsu FDSS 7000 System Pack of 19200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15581383 Supplier Axygen™ Supplier No. FDSS1536BKR
Only null left
Add to basket
Add to basket
For Hamamatsu FDSS series systems
  • For 384-Well plates
  • Tip color: Black
  • Capacity: 50μL
  • Packaging: 384 Tips/rack; 10 Racks/unit; 5 Units/Cs.
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 50 μL
Filtered Non-filtered
Sterility Non-sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance Non-ANSI
Format Rack
Tip Style Automation, Round
Packaging SBS Rack
Quantity 50 Racks/Cs.
For Use With (Equipment) Hamamatsu FDSS 7000 System
Material Polypropylene