Learn More
Epimedin A1, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Epimedin A1 is a flavonoid extracted from Herba Epimedii which is one of commonly used Chinese medicines.
Specifications
Specifications
| Chemical Name or Material | Epimedin A1 |
| CAS | 140147-77-9 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C39H50O20 |
| Quantity | 5 mg |
| Synonym | Hexandraside F |
| Solubility Information | DMSO : 100 mg/mL (119.22 mM; Need ultrasonic) |
| SMILES | O[C@H]1[C@H](O)[C@@H](O)[C@H](OC2=CC(O)=C(C(C(O[C@H]3[C@H](O)[C@H](O[C@@]4([H])[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O4)[C@@H](O)[C@H](C)O3)=C(C5=CC=C(OC)C=C5)O6)=O)C6=C2C/C=C(C)/C)O[C@@H]1CO |
| Molecular Weight (g/mol) | 838.8 |
| Show More |
Research purposes only