Learn More
Invitrogen™ PRRT1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100309
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PRRT1.
PRRT1 is a protein coding gene.
Specifications
| PRRT1 | |
| Polyclonal | |
| Unconjugated | |
| PRRT1 | |
| Bat4; Bat-4; C6orf31; C7H6orf31; D17H6S54E; Dispanin subfamily D member 1; DSPD1; G5b; IFITMD7; interferon induced transmembrane protein domain containing 7; Ng5; Orf31; proline rich transmembrane protein 1; proline-rich transmembrane protein 1; PRRT1; prrt1 {ECO:0000250; Synapse differentiation-induced protein 4; SynDIG4; UniProtKB:Q99946} | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 260297, 406167, 80863 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35449, Q6MG82, Q99946 | |
| PRRT1 | |
| A synthesized peptide derived from human PRRT1(Accession Q99946), corresponding to amino acid residues M1-S39. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |