Learn More
Human VSIG8 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB9418-100
 View more versions of this product
Product Code. 18752959
  • € 379.00 / 100µg
Description
VSIG8 Monoclonal specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry, ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VSIG8
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
V-set and immunoglobulin domain containing 8
Human embryonic kidney cell line HEK293-derived transfected with human VSIG8 Met1-Gly263 Accession # Q5VU13
100 μg
Primary
Detects human VSIG8 in direct ELISAs. Stains human VSIG8 transfectants but not irrelevant transfectants in flow cytometry.
Human
IgG1
Flow Cytometry, ELISA
961823
Flow Cytometry 0.25 ug/10^6 cells, ELISA
Q5VU13
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
391123
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.