Learn More
Human ATM Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB22901-SP
Product Code. 18761497
  • € 184.00 / 25µg
Description
ATM Monoclonal specifically detects ATM in Human samples. It is validated for Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ATM
Monoclonal
LYOPH
Immunocytochemistry 8-25 ug/mL
Q13315
ATM
E. coli-derived recombinant human ATM Arg2138-Arg2400 Accession # Q13315
25 μg
Primary
Detects human ATM in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human ATR is observed.
Human
Purified
Immunocytochemistry
664703
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AT mutated, A-T mutated, AT1, ATA, ataxia telangiectasia mutated, ataxia telangiectasia mutated (includes complementation groups A, C and D), ATC, ATD, ATDC, ATE, ATM serine/threonine kinase, DKFZp781A0353, EC 2.7.11.1, MGC74674, serine-protein kinase ATM, TEL1, TEL1, telomere maintenance 1, homolog, TELO1, TPLL
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
472
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b