Learn More
RBPJ Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30044-1-AP-20UL
This item is not returnable.
View return policy
Description
The RBPJ antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RBPJ. This antibody recognizes human, mouse antigen. The RBPJ antibody has been validated for the following applications: WB, IP, IHC, ELISA analysis.Specifications
| RBPJ | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 3516 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
| 450 μg/mL | |
| CBF1; IGKJRB; IGKJRB1; KBF2; MGC61669; RBP-J; RBPJK; RBPSUH; SUH; csl | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| ∼60 kDa | |
| Liquid |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur