Learn More
Vildagliptin, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Vildagliptin
Specifications
Specifications
| Chemical Name or Material | Vildagliptin |
| CAS | 274901-16-5 |
| Analyte or Component Names | Vildagliptin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H25 N3 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | (2S)-1-[2-[(3-Hydroxytricyclo[3.3.1.13,7]dec-1-yl)amino]acetyl]-2-pyrrolidinecarbonitrile, 2-Pyrrolidinecarbonitrile, 1-[[(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)amino]acetyl]-, (2S)- (9CI), (S)-1-[2-[(3-Hydroxyadamant-1-yl)amino]acetyl]pyrrolidine-2-carbonitrile, DSP 7238, Equa, Galvus, LAF 237, NVP-LAF 237, Vildagliptin |
| InChI Formula | InChI=1S/C17H25N3O2/c18-9-14-2-1-3-20(14)15(21)10-19-16-5-12-4-13(6-16)8-17(22,7-12)11-16/h12-14,19,22H,1-8,10-11H2/t12?,13?,14-,16?,17?/m0/s1 |
| SMILES | OC12CC3CC(C1)CC(C3)(C2)NCC(=O)N4CCC[C@H]4C#N |
| IUPAC Name | (2S)-1-[2-[(3-hydroxy-1-adamantyl)amino]acetyl]pyrrolidine-2-carbonitrile |
| Show More |