missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin 1 Antibody (3E5G11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 232.00 - € 578.00
Specifications
| Antigen | Villin 1 |
|---|---|
| Clone | 3E5G11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18082876
|
Novus Biologicals
NBP2-52585 |
0.1 mg |
€ 578.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18632648
|
Novus Biologicals
NBP2-52585-0.025mg |
0.025 mg |
€ 232.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Villin 1 Monoclonal specifically detects Villin 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| Villin 1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cytoskeleton Markers | |
| PBS with 0.05% Sodium Azide | |
| 7429 | |
| Purified recombinant fragment of human VIL1 (AA: 1-209) expressed in E. Coli. | |
| Primary | |
| 92.7 kDa |
| 3E5G11 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| D2S1471, VIL, villin 1, villin-1 | |
| VIL1 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title