missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin 1 Antibody (VIL1/1314 + VIL1/2376), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08919H
This item is not returnable.
View return policy
Description
Villin 1 Monoclonal antibody specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Villin 1 | |
| Monoclonal | |
| HRP | |
| D2S1471, VIL, villin 1, villin-1 | |
| A recombinant fragment (around aa179-311) of human Villin 1 protein (exact sequence is proprietary) (Uniprot: P09327) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 7429 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| VIL1/1314 + VIL1/2376 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido