missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Villin 1 Antibody (VIL1/8105R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21089CL1
This item is not returnable.
View return policy
Description
Villin 1 Monoclonal antibody specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Villin 1 | |
| Monoclonal | |
| CoraFluor 1 | |
| D2S1471, VIL, villin 1, villin-1 | |
| Recombinant fragment (around aa600-700) of human Villin 1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers | |
| 7429 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| VIL1/8105R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction