missing translation for 'onlineSavingsMsg'
Learn More

Villin 1 Antibody (VIL1/8105R), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-21089R

Product Code. 18568336

  • € 726.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Villin 1 Monoclonal antibody specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Villin 1
Monoclonal
DyLight 550
D2S1471, VIL, villin 1, villin-1
Recombinant fragment (around aa600-700) of human Villin 1 protein (exact sequence is proprietary)
0.1 mL
Cancer, Cell Biology, Cellular Markers
7429
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
VIL1/8105R
50 mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.