missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Villin Monoclonal Antibody (3E5G11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538658
This item is not returnable.
View return policy
Description
This antibody has been tested in indirect ELISA.
Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.
Specifications
| Villin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P09327 | |
| VIL1 | |
| Purified recombinant fragment of human VIL1 (aa: 1-209) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3E5G11 | |
| Unconjugated | |
| VIL1 | |
| D2S1471; OTTHUMP00000207159; Vil; Vil1; villin 1; villin-1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 7429 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction