missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIM Monoclonal Antibody (OTI11C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801201S
This item is not returnable.
View return policy
Description
VIM Monoclonal antibody specifically detects VIM in Human samples. It is validated for Western BlotSpecifications
| VIM | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UM73 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 238 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI11C11 | |
| Unconjugated | |
| ALK | |
| cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus | |
| Human recombinant protein fragment corresponding to amino acids 1300-1620 of human ALK produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction