missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIM Monoclonal Antibody (OTI11C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801201
This item is not returnable.
View return policy
Description
Vimentin is a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.Specifications
| VIM | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P08670 | |
| VIM | |
| Human recombit protein fragment corresponding to amino acids 210-466 of human VIM produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI11C11 | |
| Unconjugated | |
| VIM | |
| vimentin | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7431 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction