missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VIM Mouse anti-Human, Clone: VM1170, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14648.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against full length recombinant human VIM.
- High specificity
- High reactivity
Specifications
| VIM | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) Western Blot (0.25-0.5 ug/mL) The optimal working dilution should be determi | |
| P08670 | |
| VIM | |
| Recombinant protein corresponding to full length human VIM. | |
| 100 μg | |
| Primary | |
| This antibody shows no cross-reaction with other closely related intermediate filament proteins such as desmin, keratin, neurofilament, and glial fibrillary acid protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| VM1170 | |
| Mouse monoclonal antibody raised against full length recombinant human VIM. | |
| In PBS. | |
| FLJ36605 | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 7431 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction