missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vimentin Antibody (2A52), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-92688CL1
This item is not returnable.
View return policy
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human,Mouse (Negative),Rat samples. It is validated for Immunohistochemistry,Western Blot,Gene Knock-Out,Immunocytochemistry/Immunofluorescence
Specifications
| Vimentin | |
| Monoclonal | |
| CoraFluor 1 | |
| epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin | |
| Full length recombinant human Vimentin Antibody protein expressed in and purified from E. coli. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells | |
| 7431 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Gene Knock-Out, Immunocytochemistry/Immunofluorescence | |
| 2A52 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative), Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction